Learn More
Bellco Glass, Inc. Silicone Tubing 25', 5/32OD Packs of 25'
SDP
Catalog No. 501946262
Encompass_Preferred
Silicone Tubing 5/32". Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range of performance requirements, extended life pumping and in applications demanding ultimate purity in fluid transfer and peristaltic pump. Peroxide silcone tubingod: 5/32" (4.0Mm)id: 3/32" (1.6Mm)wall: 1/32" (0.8Mm) max.Pressure @ 70°c: 14packed 25' rollapolypropylenelication: pharmaceutical, biotechnology, food & beverage aerospace & general lab apolypropylenelications. Characteristics: peroxide-curred - excellent heat resistance, lowcompression set, odorless, non-toxic - translucentresin cerfication: resin is fda-compliant (21 cfr 177.2600)Meets usp class vi requirements. Tubing is manufactured to gmp temperature range: -60°f to 460°f (-51°c to 238°c)sterilize: with autoclave or gamma irradiation. Size:5/32” x 25' .
Catalog No. 50-194-6262 Supplier Bellco Glass, Inc. Supplier No. 780010400
Silicone Tubing 5/32". Silicone Tubing is specifically formulated for use with media found in pharmaceutical and biotech processing. Ideal for a range of performance requirements, extended life pumping and in applications demanding ultimate purity in fluid transfer and peristaltic pump. Peroxide silcone tubingod: 5/32" (4.0Mm)id: 3/32" (1.6Mm)wall: 1/32" (0.8Mm) max.Pressure @ 70°c: 14packed 25' rollapolypropylenelication: pharmaceutical, biotechnology, food & beverage aerospace & general lab apolypropylenelications. Characteristics: peroxide-curred - excellent heat resistance, lowcompression set, odorless, non-toxic - translucentresin cerfication: resin is fda-compliant (21 cfr 177.2600)Meets usp class vi requirements. Tubing is manufactured to gmp temperature range: -60°f to 460°f (-51°c to 238°c)sterilize: with autoclave or gamma irradiation. Size:5/32” x 25' .