Learn More
AVANTOR MASTERFLEX (VWR LLC) Peroxide-Cured Silicone Tubing, 3/8""ID x 1/2""OD, 25 ft/pack
Catalog No. NC1408915
Encompass
Peroxide-Cured Silicone Tubing, 3/8"ID x 1/2"OD, 25 ft/Pk, Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications.Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent.Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP.Temperature range: ¿60 to 460¿F (¿51 to 238¿C)Sterilize: Autoclave or gamma irradiation.
Catalog No. NC1408915 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641112
Peroxide-Cured Silicone Tubing, 3/8"ID x 1/2"OD, 25 ft/Pk, Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications.Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent.Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP.Temperature range: ¿60 to 460¿F (¿51 to 238¿C)Sterilize: Autoclave or gamma irradiation.