Learn More
Bellco Glass, Inc. Silicone TUBING, 5/16 OD Packed 25 ft lengths
SDP
Catalog No. 501946257
Encompass_Preferred
Silicone Tubing is specifically formulated for use with media foundin pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Clear silicone tubing wallcut to 25' lengths, temperature range: -51°c to 238°cmax.Pressure: 13psig @ 21°cfda compliant - 21 cfr 177.2600Meets usp class vi. Size:3/16 x 25'
Catalog No. 50-194-6257 Supplier Bellco Glass, Inc. Supplier No. 780010025
Silicone Tubing is specifically formulated for use with media foundin pharmaceutical and biotech processing. Ideal for a range ofperformance requirements, extended life pumping and in applicationsdemanding ultimate purity in fluid transfer and peristaltic pump. Clear silicone tubing wallcut to 25' lengths, temperature range: -51°c to 238°cmax.Pressure: 13psig @ 21°cfda compliant - 21 cfr 177.2600Meets usp class vi. Size:3/16 x 25'