Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Silicone Tubing, 6/9 mm
SDP

Catalog No. 05405013 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Condenser, Recirculating Chiller
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-405-013 Condenser, Recirculating Chiller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-405-013 Supplier BUCHI Supplier No. 004133
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Transparent silicone tubing with an inner diameter of 6 mm and an outer diameter of 9 mm, for use with cooling media.

  • For use with cooling media
  • Transparent silicone

Specifications

Color Transparent
Material Silicone
Type Hose
For Use With (Application) Cooling Media
For Use With (Equipment) Condenser, Recirculating Chiller
Diameter (Metric) 6 mm (Inner), 9 mm (Outer)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.