Learn More
Across International Ai 1/2" Insulated Chiller Translucent Silicone Tubing - 6 Ft
SDP
Catalog No. NC2131577
Encompass_Preferred
Ai 1/2" insulated chiller silicone tubing. Inner tubing comes in translucent natural color for visual contact with the flow, able to resist temperature from -60C to 100C. Silicon tubing ID: 5/16", OD: 11/16" Insulation ID: 1/2", OD: 1.2"
This item is not returnable. View return policy
Catalog No. NC2131577 Supplier Across International Supplier No. CT6F
Ai 1/2" insulated chiller silicone tubing. Inner tubing comes in translucent natural color for visual contact with the flow, able to resist temperature from -60C to 100C. Silicon tubing ID: 5/16", OD: 11/16" Insulation ID: 1/2", OD: 1.2"