Learn More
WESTLAB Silicone UNI-STOP Stopper - Small 10pk
Catalog No. NC3825887
Encompass
The Westlab ‘UNI-STOP’ stopper is an innovative product designed by Westlab to replace all of your stoppers. Each stopper has the ability to transform from a solid stopper to a stopper with one hole for a range of experiments.Reverse grip-ribs for maximum seal.Two in one, solid or one hole.Universal, fits multiple neck sizes.Internal barb seal, for ease of tubing insert.Universal hole, fits tubing from 6-10mm D.A truly innovative product, unique to Westlab.
Catalog No. NC3825887 Supplier WESTLAB Supplier No. 663393C
The Westlab ‘UNI-STOP’ stopper is an innovative product designed by Westlab to replace all of your stoppers. Each stopper has the ability to transform from a solid stopper to a stopper with one hole for a range of experiments.Reverse grip-ribs for maximum seal.Two in one, solid or one hole.Universal, fits multiple neck sizes.Internal barb seal, for ease of tubing insert.Universal hole, fits tubing from 6-10mm D.A truly innovative product, unique to Westlab.