Learn More
Anton Paar SILICONE VISCOSITY STANDARD AP RT30k600 ml - 33 / 30 Pa.s at 20 / 25 °CQT# 820262286
Catalog No. NC2894799
Encompass_Preferred
SILICONE VISCOSITY STANDARD AP RT30k600 ml - 33 / 30 Pa.s at 20 / 25 °CQT# 820262286
Catalog No. NC2894799 Supplier Anton Paar Supplier No. 191120
SILICONE VISCOSITY STANDARD AP RT30k600 ml - 33 / 30 Pa.s at 20 / 25 °CQT# 820262286