Learn More
AVANTOR MASTERFLEX (VWR LLC) Silicone w/SS Perforated Plate; 3/4"" Tri-Clamp
Catalog No. NC1656947
Encompass
Gauge Guard Protectors safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals. Gauge Guard Isolator Seals combine a hygenic seal with a membrane to isolate hot or caustic solutions from your instrument. Available in platinum-cured silicone or EPDM.
Catalog No. NC1656947 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3056501
Gauge Guard Protectors safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals. Gauge Guard Isolator Seals combine a hygenic seal with a membrane to isolate hot or caustic solutions from your instrument. Available in platinum-cured silicone or EPDM.