Learn More
Active Motif Siliconized Tubes, 1.7 ml
SDP
Catalog No. NC1852562
Encompass
Because polystyrene transfers sonic wave energy much more efficiently than polypropylene, Active Motif highly recommends that its Polystyrene Sonication Tubes be used with its Q800R Sonicator. Use of polypropylene tubes in indirect sonication has been shown to reduce the extent to which the chromatin is sheared
Catalog No. NC1852562 Supplier Active Motif Supplier No. 53036
Because polystyrene transfers sonic wave energy much more efficiently than polypropylene, Active Motif highly recommends that its Polystyrene Sonication Tubes be used with its Q800R Sonicator. Use of polypropylene tubes in indirect sonication has been shown to reduce the extent to which the chromatin is sheared