Learn More
LabScientific Silane Coated Microscope slides. Clear, white, frosted edge. Clean Room prepared. Charged. Allows for covalent bonding of tissue sections and cytology preparations to glass slide. 1"x3". 72/pack.
Catalog No. NC9968319
Encompass
Silane Coated Microscope Slides, 3-Aminopropyltriethoxysilane,RNase Free,Positively Charged,Clear with White Frosted Edge,Clean Room Prepared,allow covalent bonding of tissue sections and cytology preparations to glass slides, 72 Slides/Box
Catalog No. NC9968319 Supplier LabScientific Supplier No. 7801
Silane Coated Microscope Slides, 3-Aminopropyltriethoxysilane,RNase Free,Positively Charged,Clear with White Frosted Edge,Clean Room Prepared,allow covalent bonding of tissue sections and cytology preparations to glass slides, 72 Slides/Box