Learn More
Bollé Safety Silium+ Safety Glasses
Catalog No. 18072108
Change view
Click to view available options
Lens Tint:
CSP
Clear
Smoke
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
18-072-108 Smoke
18-072-106 Clear
18-072-107 CSP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-072-108 Supplier Bollé Safety Supplier No. 40238
Only null left
Add to Cart
Add to Cart
Silium+ safety glasses are comfortable, ultra-lightweight combining style, ergonomics and performances for extended use
  • Ultra-lightweight frame
  • All-round vision
  • Polycarbonate lens
  • Ergonomic flexible temples
  • Adjustable non-slip bridge
  • Flexible, thin temple arms
  • 99.99% UVA and UVB light absorption
Specifications
Lens Coating Anti-fog, Scratch-resistant
Shape Rectangular
Certifications/Compliance ANSI Z87+
Lens Tint Smoke
Product Type Safety Glasses
Frame Color Gunmetal
Weight (English) 0.74 oz.
Material Polycarbonate,TPR
Disposable Reusable
Lens Material Polycarbonate
Frame Material Gunmetal
UV Protection UVA and UVB
Frame Type Half Frame