Learn More
Electron Microscopy Sciences Silk, P/B, N/H, 10"
SDP
Catalog No. 50997219
Encompass_Preferred
Made of 100% pure woven silk, these polishing cloths are recommended for intermediate, final, or skid polishing on friable materials. The cloths have a plain back and No Hole.
Type: no hole
Diameter: 10"
Catalog No. 50-997-219 Supplier Electron Microscopy Sciences Supplier No. 5031205
Made of 100% pure woven silk, these polishing cloths are recommended for intermediate, final, or skid polishing on friable materials. The cloths have a plain back and No Hole.
Type: no hole
Diameter: 10"