Learn More
Grainger Approved Vendor Sillcock, Quarter-Turn, Brass, 1/2 in. FNPT, 1 ea.
Catalog No. 502793360
Encompass_Preferred
This angle sillcock is an outdoor faucet designed to provide convenient access to water for applications such as washing vehicles or gardening. Featuring a durable brass construction and a chrome ball disc, this unit ensures reliable flow control in a standard design.
  • 1/2 in. FNPT inlet connection size
  • 3/4 in. GHT outlet connection
  • Quarter-turn handle operation
  • 125 psi maximum pressure
  • Compliant with ANSI B1.20.1, ANSI B1.20.7, ANSI/NSF 61-9, and ASME A112.18.1 standards
Catalog No. 50-279-3360 Supplier Grainger Supplier No. 1APY4
This angle sillcock is an outdoor faucet designed to provide convenient access to water for applications such as washing vehicles or gardening. Featuring a durable brass construction and a chrome ball disc, this unit ensures reliable flow control in a standard design.
  • 1/2 in. FNPT inlet connection size
  • 3/4 in. GHT outlet connection
  • Quarter-turn handle operation
  • 125 psi maximum pressure
  • Compliant with ANSI B1.20.1, ANSI B1.20.7, ANSI/NSF 61-9, and ASME A112.18.1 standards