Learn More
Grainger Zoro Select Sillcock, Brass, 3/4 in FNPT, Quarter-Turn, 125 psi
Catalog No. 502793361
Encompass_Preferred
This angle sillcock is a durable brass valve designed for non-potable water applications. Featuring a convenient quarter-turn handle, it provides reliable flow control for irrigation or general outdoor utility systems.
  • Body material: brass
  • Handle material: aluminum
  • Inlet size: 3/4 in. FNPT
  • Outlet connection type: GHT
  • Maximum pressure: 125 psi
  • Design: not frost-resistant
  • Standards met: ANSI B1.20.1, ANSI B1.20.7, and ASME A112.18.1/CSA B125.1
  • Warning: this product is not intended for potable water applications and contains materials known to the State of California to cause cancer and birth defects or other reproductive harm
Catalog No. 50-279-3361 Supplier Grainger Supplier No. 1APY5
This angle sillcock is a durable brass valve designed for non-potable water applications. Featuring a convenient quarter-turn handle, it provides reliable flow control for irrigation or general outdoor utility systems.
  • Body material: brass
  • Handle material: aluminum
  • Inlet size: 3/4 in. FNPT
  • Outlet connection type: GHT
  • Maximum pressure: 125 psi
  • Design: not frost-resistant
  • Standards met: ANSI B1.20.1, ANSI B1.20.7, and ASME A112.18.1/CSA B125.1
  • Warning: this product is not intended for potable water applications and contains materials known to the State of California to cause cancer and birth defects or other reproductive harm