Learn More
Selleck Chemical LLC Silmitasertib (CX-4945) S2248-50mg
SDP
Catalog No. 503111868
Encompass_Preferred
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay less potent to Flt3 Pim1 and CDK1 (inactive in cell-based assay) Silmitasertib induces autophagy and promotes apoptosis Phase 1/2
Catalog No. 50-311-1868 Supplier Selleck Chemical LLC Supplier No. S224850MG
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay less potent to Flt3 Pim1 and CDK1 (inactive in cell-based assay) Silmitasertib induces autophagy and promotes apoptosis Phase 1/2