Learn More
Selleck Chemical LLC Silmitasertib (CX-4945) 2mg 1009820-21-6
SDP
Catalog No. NC1353071
Encompass_Preferred
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay, less potent to Flt3, Pim1 and CDK1 (inactive in cell-based assay). Silmitasertib induces autophagy and promotes apoptosis. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1353071 Supplier Selleck Chemical LLC Supplier No. S22482MG
Add to Cart
Add to Cart
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay, less potent to Flt3, Pim1 and CDK1 (inactive in cell-based assay). Silmitasertib induces autophagy and promotes apoptosis. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.