Learn More
Medchemexpress LLC HY-50855B 10mg Medchemexpress, Silmitasertib (sodium salt) CAS:1309357-15-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50855B10MG
Encompass_Preferred
Medchemexpress, HY-50855B 10mg Silmitasertib (sodium salt) CAS:1309357-15-0 Silmitasertib sodium salt is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9340
 Restricted Item