Learn More
TARGETMOL CHEMICALS INC SILMITASERTIB SODIUM SAL 25MG
SDP
Catalog No. 502581258
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Silmitasertib sodium salt (CX-4945 sodium salt) is a potent and orally bioavailable highly selective inhibitor of CK2(IC50 of 1 nM CK2(alpha)). purity: 99%
Catalog No. 50-258-1258 Supplier TARGETMOL CHEMICALS INC Supplier No. T1502425MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Silmitasertib sodium salt (CX-4945 sodium salt) is a potent and orally bioavailable highly selective inhibitor of CK2(IC50 of 1 nM CK2(alpha)). purity: 99%