Learn More
Medchemexpress LLC Silmitasertib sodium salt | 1309357-15-0 | 99.9% | C19H11ClN3NaO2 | 100 MG
SDP
Catalog No. 5000357913
Encompass_Preferred
Silmitasertib sodium salt is an orally bioavailable, highly selective and potent CK2 inhibitor.
  • Causes cell-cycle arrest and selectively induces apoptosis in cancer cells by attenuating PI3K/Akt signaling.
  • Antiproliferative activity correlates with CK2α catalytic subunit expression.
  • Prevents leukemic cells from engaging UPR during PS-341 treatment, reducing BIP/Grp78 expression.
  • Induces cytotoxicity and apoptosis, exerting anti-proliferative effects in hematological tumors.
  • Demonstrates robust antitumor activity and reduces phospho-p21 (T145) in murine xenograft models.
Catalog No. 50-003-57913 Supplier Medchemexpress LLC Supplier No. HY50855B100MG
Silmitasertib sodium salt is an orally bioavailable, highly selective and potent CK2 inhibitor.
  • Causes cell-cycle arrest and selectively induces apoptosis in cancer cells by attenuating PI3K/Akt signaling.
  • Antiproliferative activity correlates with CK2α catalytic subunit expression.
  • Prevents leukemic cells from engaging UPR during PS-341 treatment, reducing BIP/Grp78 expression.
  • Induces cytotoxicity and apoptosis, exerting anti-proliferative effects in hematological tumors.
  • Demonstrates robust antitumor activity and reduces phospho-p21 (T145) in murine xenograft models.