Learn More
Medchemexpress LLC Silmitasertib sodium salt | 1309357-15-0 | 99.9% | C19H11ClN3NaO2 | 5 MG
SDP
Catalog No. 5000358086
Encompass_Preferred
Silmitasertib sodium salt is an orally bioavailable, highly selective, and potent CK2 inhibitor (IC50 values of 1 nM against CK2α and CK2α'). It induces cell-cycle arrest and selectively causes apoptosis in cancer cells, attenuates PI3K/Akt signaling, and demonstrates antiproliferative activity correlating with CK2α catalytic subunit expression. It also prevents leukemic cells from engaging a functional UPR to buffer proteotoxic stress and decreases pro-survival ER chaperon BIP/Grp78 expression.
  • Causes cell-cycle arrest
  • Induces apoptosis in cancer cells
  • Attenuates PI3K/Akt signaling
  • Exhibits antiproliferative activity
  • Prevents leukemic cells from engaging a functional UPR
  • Decreases pro-survival ER chaperon BIP/Grp78 expression
  • Induces cytotoxicity
  • Suppresses CK2-mediated PI3K/Akt/mTOR signaling pathways
  • Demonstrates robust antitumor activity in murine xenograft models
Catalog No. 50-003-58086 Supplier Medchemexpress LLC Supplier No. HY50855B5MG
Silmitasertib sodium salt is an orally bioavailable, highly selective, and potent CK2 inhibitor (IC50 values of 1 nM against CK2α and CK2α'). It induces cell-cycle arrest and selectively causes apoptosis in cancer cells, attenuates PI3K/Akt signaling, and demonstrates antiproliferative activity correlating with CK2α catalytic subunit expression. It also prevents leukemic cells from engaging a functional UPR to buffer proteotoxic stress and decreases pro-survival ER chaperon BIP/Grp78 expression.
  • Causes cell-cycle arrest
  • Induces apoptosis in cancer cells
  • Attenuates PI3K/Akt signaling
  • Exhibits antiproliferative activity
  • Prevents leukemic cells from engaging a functional UPR
  • Decreases pro-survival ER chaperon BIP/Grp78 expression
  • Induces cytotoxicity
  • Suppresses CK2-mediated PI3K/Akt/mTOR signaling pathways
  • Demonstrates robust antitumor activity in murine xenograft models