Learn More
Medchemexpress LLC Silmitasertib sodium salt | 1309357-15-0 | 99.9% | C19H11ClN3NaO2 | 500 MG
SDP
Catalog No. 5000358089
Encompass_Preferred
Silmitasertib sodium salt is an orally bioavailable, highly selective, and potent CK2 inhibitor. It induces cell-cycle arrest and selective apoptosis in cancer cells, attenuating PI3K/Akt signaling.
  • Orally bioavailable
  • Highly selective and potent CK2 inhibitor
  • Induces cell-cycle arrest in cancer cells
  • Selectively induces apoptosis in cancer cells
  • Attenuates PI3K/Akt signaling
  • Demonstrates robust antitumor activity in murine xenograft models
  • Well-tolerated in vivo
Catalog No. 50-003-58089 Supplier Medchemexpress LLC Supplier No. HY50855B500MG
Silmitasertib sodium salt is an orally bioavailable, highly selective, and potent CK2 inhibitor. It induces cell-cycle arrest and selective apoptosis in cancer cells, attenuating PI3K/Akt signaling.
  • Orally bioavailable
  • Highly selective and potent CK2 inhibitor
  • Induces cell-cycle arrest in cancer cells
  • Selectively induces apoptosis in cancer cells
  • Attenuates PI3K/Akt signaling
  • Demonstrates robust antitumor activity in murine xenograft models
  • Well-tolerated in vivo