Learn More
Medchemexpress LLC Silmitasertib sodium salt (CX-4945 sodium salt) | 1309357-15-0 | 99.9% | C19H11ClN3NaO2 | 50 MG
SDP
Catalog No. 5000358773
Encompass_Preferred
Silmitasertib sodium salt is an orally bioavailable, highly selective, and potent CK2 inhibitor, demonstrating IC50 values of 1 nM against CK2α and CK2α'. This compound induces cell-cycle arrest and selectively promotes apoptosis in cancer cells. It also attenuates PI3K/Akt signaling, decreases pro-survival ER chaperon BIP/Grp78 expression, and suppresses activation of CK2-mediated PI3K/Akt/mTOR signaling pathways in hematological tumors.
  • Orally bioavailable and potent CK2 inhibitor
  • Highly selective against CK2α and CK2α' (IC50 1 nM)
  • Induces cell-cycle arrest and apoptosis in cancer cells
  • Attenuates PI3K/Akt signaling
  • Suppresses CK2-mediated PI3K/Akt/mTOR signaling
Catalog No. 50-003-58773 Supplier Medchemexpress LLC Supplier No. HY50855B50MG
Silmitasertib sodium salt is an orally bioavailable, highly selective, and potent CK2 inhibitor, demonstrating IC50 values of 1 nM against CK2α and CK2α'. This compound induces cell-cycle arrest and selectively promotes apoptosis in cancer cells. It also attenuates PI3K/Akt signaling, decreases pro-survival ER chaperon BIP/Grp78 expression, and suppresses activation of CK2-mediated PI3K/Akt/mTOR signaling pathways in hematological tumors.
  • Orally bioavailable and potent CK2 inhibitor
  • Highly selective against CK2α and CK2α' (IC50 1 nM)
  • Induces cell-cycle arrest and apoptosis in cancer cells
  • Attenuates PI3K/Akt signaling
  • Suppresses CK2-mediated PI3K/Akt/mTOR signaling