Learn More
Selleck Chemical LLC Silodosin 5mg 160970-54-7 KAD 3213, KMD 3213
SDP
Supplier:  Selleck Chemical LLC S16135MG
Encompass_Preferred
Silodosin (KAD 3213, KMD 3213) is a highly selective ?1A-adrenoceptor antagonist, used in treatment of benign prostatic hyperplasia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1429