Learn More
Cayman Chemical Silodosin-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1426173-86-5, 1 mg
SDP
Catalog No. 502989384
Encompass_Preferred
An internal standard for the quantification of silodosin by GC- or LC-MS
Catalog No. 50-298-9384 Supplier Cayman Chemical Supplier No. 302361 MG
An internal standard for the quantification of silodosin by GC- or LC-MS