Learn More
BURKLE INC SiloDrill, V2A, 59.06 in. (150 cm), spiral screw Ø 3.54 in. (90 mm), 13.53 oz. (400 ml)
Supplier:  BURKLE INC 53012000
Encompass_Preferred
The SiloDrill is a sampler specifically designed for taking samples of bulk materials from silos or tanks.
When bulk materials are transported over long distances, vibration and movement during transport can cause segregation of material layers, making representative sampling difficult.
The SiloDrill takes an all-layer sample through multiple layers of the bulk material so that the quality, composition, or other relevant properties of the entire stored material can be analyzed.
The SiloDrill fits through openings as small as 90 mm in diameter, making it very versatile. A special mechanism makes it easy to open and close the sampling chamber by turning the rugged T-handle. The spring-loaded inner tube minimizes the risk of jamming coarse grains. Drilling paddles and a scraping edge also allow sampling of caked and non-flowing bulk materials.
Catalog No. 50-000-25828
Add to Cart
Request a Quote