Learn More
Huber USA Heat transfer fluid M20.195/235.20 | 5L
Catalog No. NC1780509
Encompass_Preferred
Temperature range for: - open/closed systems: -20°C…195°C - Unistats (only closed): -20°C…235°C | Material: silicon oil | Colour: colourless, clear | Viscosity at 25°C: 20 mm²/s
Catalog No. NC1780509 Supplier Huber USA Supplier No. 6161
Temperature range for: - open/closed systems: -20°C…195°C - Unistats (only closed): -20°C…235°C | Material: silicon oil | Colour: colourless, clear | Viscosity at 25°C: 20 mm²/s