Learn More
Sorbent Technologies Inc Silica PF - Prep TLC, 60A, w/UV254, 5-50um, 1kg
SDP
Catalog No. 501138725
Encompass_Preferred
Silica Gel for Prep TLC with UV254 fluorescent indicator has a pore size of 60A, particle distribution of 5-50um, and a quantity of 1kg. When combined with a binding agent, this silica gel can be used for the making of preparative TLC plates.
Catalog No. 50-113-8725 Supplier Sorbent Technologies Inc Supplier No. 116501
Silica Gel for Prep TLC with UV254 fluorescent indicator has a pore size of 60A, particle distribution of 5-50um, and a quantity of 1kg. When combined with a binding agent, this silica gel can be used for the making of preparative TLC plates.