Learn More
AdipoGen Silthiopham
SDP
Supplier:  AdipoGen CDXS0049M025
Encompass_Preferred
Chemical. CAS 175217-20-6. Formula C13H21NOSSi. MW 267.5. Synthetic. Amide fungicide. Interferes with fungal respiration by inhibition of ATP-Production in mitochondrias. Compound can be used as analytical reference material.
Catalog No. 50-256-0080