Learn More
Trajan Scientific SilTite Ferrule Medical Lab 10/PK
SDP
Catalog No. 5000493578
Encompass_Preferred
Replacement SilTite metal ferrules for all Thermo Scientific GCMS interface connections for use with 1/32 columns PK 10
Catalog No. 50-004-93578 Supplier Trajan Scientific Supplier No. 073219
Replacement SilTite metal ferrules for all Thermo Scientific GCMS interface connections for use with 1/32 columns PK 10