Learn More
Trajan Scientific SilTite Ferrule Medical Lab 10/PK
SDP
Supplier:  Trajan Scientific 073222
Encompass_Preferred
Replacement SilTite metal ferrules for all Thermo Scientific GCMS interface connections for use with 0 45-0 53 mm ID columns PK 10
Catalog No. 50-004-94409