Learn More
Trajan Scientific SilTite Ferrule Medical Lab 10/PK
Supplier: Trajan Scientific 073221
Replacement SilTite metal ferrules for all Thermo Scientific GCMS interface connections for use with 0 32 mm ID columns PK 10