Learn More
Trajan Scientific SilTite Ferrule Mini Medical 10/PK
SDP
Supplier:  Trajan Scientific 073471
Encompass_Preferred
Replacement SilTite mini union ferrules for 0 32 mm ID columns/fused silica PK10
Catalog No. 50-004-94905