Learn More
Trajan Scientific SilTite Ferrule Mini Medical 10/PK
SDP
Catalog No. 5000493596
Encompass_Preferred
Replacement SilTite mini union ferrules for 0 1 - 0 25 mm ID columns/fused silica PK10
Catalog No. 50-004-93596 Supplier Trajan Scientific Supplier No. 073470
Replacement SilTite mini union ferrules for 0 1 - 0 25 mm ID columns/fused silica PK10