Learn More
Trajan Scientific SilTite Ferrule Shimadzu Medical 10/PK
SDP
Supplier:  Trajan Scientific 073229
Encompass_Preferred
SilTite ferrules for Shimadzu GC-2014 GC-2010 GC-17A and GC-14A detector/injectors and QP5000/5050 standard MS interface (not for MS interfaces or QP2010 injector) for use with 0 45-0 53 mm ID columns PK10
Catalog No. 50-004-94305