Learn More
Trajan Scientific SilTite Ferrule Shimadzu Medical 10/PK
SDP
Supplier:  Trajan Scientific 073351
Encompass_Preferred
SilTite metal - initial installation for Shimadzu GC-2014 GC-2010 GC-17A and GC-14A detector/injectors and QP5000/5050 standard MS interface (not for MS interfaces or QP2010 injector) for 0 32 mm ID columns (10 ferrules 2 nuts)
Catalog No. 50-004-94327