Learn More
Trajan Scientific SilTite Ferrule Thermo Medical 10/PK
SDP
Catalog No. 5000494313
Encompass_Preferred
SilTite ferrules for use with 0 45-0 53 mm ID columns PK10
Catalog No. 50-004-94313 Supplier Trajan Scientific Supplier No. 073332
SilTite ferrules for use with 0 45-0 53 mm ID columns PK10