Learn More
Trajan Scientific SilTite FingerTite Detector FID 1/EA
SDP
Catalog No. 5000494606
Encompass_Preferred
SilTite FingerTite Agilent FID adaptor (includes 2 graphite Vespel FID ferrules)
Catalog No. 50-004-94606 Supplier Trajan Scientific Supplier No. 0736103
SilTite FingerTite Agilent FID adaptor (includes 2 graphite Vespel FID ferrules)