Learn More
Trajan Scientific SilTite FingerTite Injector Medical 1/EA
SDP
Catalog No. 5000493629
Encompass_Preferred
SilTite FingerTite Agilent injector adaptor (includes 2 base Seals) also suits Thermo 1300 injector
Catalog No. 50-004-93629 Supplier Trajan Scientific Supplier No. 0736104
SilTite FingerTite Agilent injector adaptor (includes 2 base Seals) also suits Thermo 1300 injector