Learn More
Trajan Scientific SilTite FingerTite Kit Agilent 1/EA
SDP
Supplier:  Trajan Scientific 07361054
Encompass_Preferred
SilTite FingerTite Agilent injector starter kit for use with 0 53 mm ID columns (10 x 0 7 mm ferrules)
Catalog No. 50-004-94900