Learn More
Trajan Scientific SilTite FingerTite Kit Agilent 1/EA
SDP
Supplier:  Trajan Scientific 07361053
Encompass_Preferred
SilTite FingerTite Agilent INJ/FID starter kit for use with 0 53 mm ID columns
Catalog No. 50-004-94939