Learn More
Trajan Scientific SilTite FingerTite Kit Shimadzu 1/EA
SDP
Supplier:  Trajan Scientific 07362053
Encompass_Preferred
SilTite FingerTite Shimadzu 2010 INJ/FID starter kit for use with 0 53 mm ID columns
Catalog No. 50-004-94648