Learn More
Trajan Scientific SilTite FingerTite Kit Starter 1/EA
SDP
Catalog No. 5000494461
Encompass_Preferred
SilTite FingerTite starter kit (5 x female nuts 10 x 0 5 mm hole ferrules 1 x measuring tool)
Catalog No. 50-004-94461 Supplier Trajan Scientific Supplier No. 0736105
SilTite FingerTite starter kit (5 x female nuts 10 x 0 5 mm hole ferrules 1 x measuring tool)