Learn More
Trajan Scientific SilTite FingerTite Kit Varian 1/PK
SDP
Catalog No. 5000494396
Encompass_Preferred
SilTite FingerTite Bruker/Varian INJ/FID starter kit
Catalog No. 50-004-94396 Supplier Trajan Scientific Supplier No. 073618
SilTite FingerTite Bruker/Varian INJ/FID starter kit