Learn More
Trajan Scientific SilTite Kit Agilent Injector 10/PK
SDP
Supplier:  Trajan Scientific 073271
Encompass_Preferred
SilTite metal - initial installation for Thermo Scientific TRACE 1300 series GC split/splitless injectors for use with 0 32 mm ID columns (10 ferrules 2 nuts 2 inlet base seals)
Catalog No. 50-004-94416