Learn More
Trajan Scientific SilTite Kit Agilent Injector 10/PK
SDP
Catalog No. 5000494395
Encompass_Preferred
SilTite metal - initial installation for Thermo Scientific TRACE 1300 series GC split/splitless injectors for use with 1/32 columns (10 ferrules 2 nuts 2 inlet base seals)
Catalog No. 50-004-94395 Supplier Trajan Scientific Supplier No. 073273
SilTite metal - initial installation for Thermo Scientific TRACE 1300 series GC split/splitless injectors for use with 1/32 columns (10 ferrules 2 nuts 2 inlet base seals)