Learn More
Trajan Scientific SilTite Kit Medical Lab 10/PK
SDP
Catalog No. 5000494108
Encompass_Preferred
SilTite metal - initial installation for GCMS interface connection for use with 0 32 mm ID columns (10 ferrules 2 nuts)
Catalog No. 50-004-94108 Supplier Trajan Scientific Supplier No. 073201
SilTite metal - initial installation for GCMS interface connection for use with 0 32 mm ID columns (10 ferrules 2 nuts)