Learn More
Trajan Scientific SilTite Kit Shimadzu Medical 10/PK
SDP
Supplier:  Trajan Scientific 073204
Encompass_Preferred
SilTite metal - initial installation for Shimadzu QP5000/5050 standard MS interface for use with 0 10-0 25 mm ID columns (10 ferrules 2 nuts)
Catalog No. 50-004-94450