Learn More
Trajan Scientific SilTite Kit Shimadzu Medical 10/PK
SDP
Catalog No. 5000494278
Encompass_Preferred
SilTite metal - initial installation for Shimadzu QP5000/5050 standard MS interface for use with 0 32 mm ID columns (10 ferrules 2 nuts)
Catalog No. 50-004-94278 Supplier Trajan Scientific Supplier No. 073205
SilTite metal - initial installation for Shimadzu QP5000/5050 standard MS interface for use with 0 32 mm ID columns (10 ferrules 2 nuts)