Learn More
Trajan Scientific SilTite Kit Thermo Finnigan 10/PK
SDP
Supplier:  Trajan Scientific 073452
Encompass_Preferred
SilTite metal - initial installation 0 45-0 53 mm ID columns (10 ferrules 2 nuts)
Catalog No. 50-004-94679