Learn More
Trajan Scientific SilTite Kit Thermo Finnigan 10/PK
SDP
Catalog No. 5000494680
Encompass_Preferred
SilTite metal - initial installation for 0 10-0 25 mm ID columns (10 ferrules 2 nuts)
Catalog No. 50-004-94680 Supplier Trajan Scientific Supplier No. 073450
SilTite metal - initial installation for 0 10-0 25 mm ID columns (10 ferrules 2 nuts)